Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Human ENA-78 (CXCL5) (8-78 aa) Recombinant Protein, PeproTech®

Catalog No. 30022B1MG
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
100 μg
20 μg
250 μg
5 μg
500 μg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
300-22B-1MG 1 mg
30022B100UG 100 μg
30022B20UG 20 μg
30022B250UG 250 μg
30022B500UG 500 μg
300-22B-5UG 5 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 300-22B-1MG Supplier Gibco™ Supplier No. 30022B1MG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

Recombinant Human ENA-78 is a 7.8 kDa protein consisting of 71 amino acid residues. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

The protein encoded by this gene is an inflammatory chemokine that belongs to the CXC chemokine family. This chemokine is produced concomitantly with interleukin-8 (IL8) in response to stimulation with either interleukin-1 (IL1) or tumor necrosis factor-alpha (TNFA). This chemokine is a potent chemotaxin involved in neutrophil activation.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P42830
For Use With (Application) Functional Assay
Formulation protein with no preservative
Gene ID (Entrez) 6374
Molecular Weight (g/mol) 7.8 kDa
Name Human ENA-78 (CXCL5) (8-78 aa)
Quantity 1 mg
Source E. coli
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias AMCF-II; C Cmotif chemokine; C x C motif chemokine; CC motif chemokine; CCmotif chemokine; chemokine (C-X-C motif) ligand 5; chemokine (C-X-C motif) ligand 6; chemokine (C-X-C motif) ligand 6 (granulocyte chemotactic protein 2); CXC; CXC chemokine LIX; CXC motif chemokine; C-X-C motif chemokine 5; C-X-C motif chemokine ligand 5; CXCL; CXCL5; Cxcl6; Cytokine LIX; ENA78; ENA-78; ENA-78(1-78); ENA-78(8-78); ENA-78(9-78); Epithelial-derived neutrophil-activating protein 78; GCP-2; GCP-2(1-78); GCP-2(9-78); granulocyte chemotactic protein-2; LIX; Neutrophil-activating peptide ENA-78; neutrophil-activating protein 78; Scyb5; Scyb6; small inducible cytokine B subfamily, member 5; small inducible cytokine subfamily B (Cys-X-Cys), member 5 (epithelial-derived neutrophil-activating peptide 78); small inducible cytokine subfamily B, member 15; small-inducible cytokine B5
Common Name CXCL5
Gene Symbol CXCL5
Biological Activity Determined by its ability to chemoattract human neutrophils using a concentration range of 10.0-100.0 ng/ml
Conjugate Unconjugated
Recombinant Recombinant
Sequence LRELRCVCLQ TTQGVHPKMI SNLQVFAIGP QCSKVEVVAS LKNGKEICLD PEAPFLKKVI QKILDGGNKE N
Content And Storage -20°C
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.