Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human ENAM Partial ORF (NP_114095, 1043 a.a. - 1141 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89943253 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-943-253 25 μg
89-943-252 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-943-253 Supplier Abnova™ Supplier No. H00010117Q01L
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

Dental enamel is a highly mineralized tissue with 85% of its volume occupied by unusually large, highly organized, hydroxyapatite crystals. This highly organized and unusual structure is thought to be rigorously controlled in ameloblasts through the interaction of a number of organic matrix molecules that include enamelin, amelogenin (AMELX; MIM 300391), ameloblastin (AMBN; MIM 601259), tuftelin (TUFT1; MIM 600087), dentine sialophosphoprotein (DSPP; MIM 125485), and a variety of enzymes. Enamelin is the largest protein in the enamel matrix of developing teeth and comprises approximately 5% of total enamel matrix protein.[supplied by OMIM]

Sequence: ERQQQRPSNILHLPCFGSKLAKHHSSTTGTPSSDGRQSPFDGDSITPTENPNTLVELATEEQFKSINVDPLDADEHSPFEFLQRGTNVQDQVQDCLLLQ

Specifications

Accession Number NP_114095
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 10117
Molecular Weight (g/mol) 36.63kDa
Name ENAM (Human) Recombinant Protein (Q01)
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 25 μg
Immunogen ERQQQRPSNILHLPCFGSKLAKHHSSTTGTPSSDGRQSPFDGDSITPTENPNTLVELATEEQFKSINVDPLDADEHSPFEFLQRGTNVQDQVQDCLLLQ
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias ADAI/AI1C/AIH2
Common Name ENAM
Gene Symbol ENAM
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.