Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human ENDOGL1 Partial ORF (NP_005098.1, 269 a.a. - 368 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89943262 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-943-262 25 μg
89-943-261 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-943-262 Supplier Abnova™ Supplier No. H00009941Q01L
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

This gene encodes an endo/exonuclease with 5'-3' exonuclease activity. The encoded enzyme catalyzes the hydrolysis of ester linkages at the 5' end of a nucleic acid chain. This enzyme is localized to the mitochondria and may play a role in programmed cell death. Alternatively spliced transcript variants have been described. A pseudogene exists on chromosome 18. [provided by RefSeq]

Sequence: LQDLEKLSGLVFFPHLDRTSDIRNICSVDTCKLLDFQEFTLYLSTRKIEGARSVLRLEKIMENLKNAEIEPDDYFMSRYEKKLEELKAKEQSGTQIRKPS

Specifications

Accession Number NP_005098.1
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 9941
Molecular Weight (g/mol) 36.74kDa
Name ENDOGL1 (Human) Recombinant Protein (Q01)
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 25 μg
Immunogen LQDLEKLSGLVFFPHLDRTSDIRNICSVDTCKLLDFQEFTLYLSTRKIEGARSVLRLEKIMENLKNAEIEPDDYFMSRYEKKLEELKAKEQSGTQIRKPS
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias ENDOGL1/ENDOGL2/ENGL/ENGL-B/ENGL-a/MGC125944/MGC125945
Common Name EXOG
Gene Symbol EXOG
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.