Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Human Endostatin Recombinant Protein, PeproTech®

Catalog No. 15001500UG
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
100 μg
20 μg
250 μg
500 μg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
15001500UG 500 μg
15001100UG 100 μg
150-01-1MG 1 mg
150-01-20UG 20 μg
15001250UG 250 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 15001500UG Supplier Gibco™ Supplier No. 15001500UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

Recombinant Human Endostatin is a 20.2 kDa protein consisting of 184 amino acid residues. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

Endostatin is able to avoke non-uniform response for proliferation, cell mount and migration of entothelial cells, which different endostatin binding characteristic, leads to the assumption that endostatin effect is strongly dependent from endothelial cell type. Furthermore endostatin inhibits angiogenesis and tumor growth in vivo by inducing apoptosis in endothelial cells. The local delivery of endostatin seems to specifically affect tumor-associated microvessels by reduction of the vessel density, diameter and functionality. Tumor cell migration and invasion was greatly reduced in the endostatin treated animals. Endostatin is non-toxic and does not induce aquired drug resistance and has therefor become a potent new therapy strategy in solid neoplasias. This therapy appears to have a high potential not only for the treatment of gliomas, the most common brain tumors, but also in other tumors. The ability of endostatin to inhibit neoangiogenesis is mediated, at least in part, by Zn^+2 binding and elastase processing. Widespread endostatin expression was found in elactic fibers in vessel walls and in some other basement membrane zones. Endostatin is released by neurons to accumulate in amyloid plaques in Alzheimer's disease.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P39060
For Use With (Application) ELISA, Functional Assay, Western Blot
Formulation protein with no preservative
Gene ID (Entrez) 80781
Molecular Weight (g/mol) 20.2 kDa
Name Human Endostatin
Quantity 500 μg
Source E. coli
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias antiangiogenic agent; Col18a1; Collagen alpha-1(XVIII) chain; collagen type XVIII alpha 1 chain; collagen, type XVIII, alpha 1; endostatin; FLJ27325; FLJ34914; KNO; KNO1; KS; MGC74745; multi-functional protein MFP; NC1; Non-collagenous domain 1; procollagen, type XVIII, alpha 1
Common Name Endostatin
Gene Symbol COL18A1
Biological Activity Measured by its ability to inhibit HUVEC migration.
Conjugate Unconjugated
Recombinant Recombinant
Sequence MHSHRDFQPV LHLVALNSPL SGGMRGIRGA DFQCFQQARA VGLAGTFRAF LSSRLQDLYS IVRRADRAAV PIVNLKDELL FPSWEALFSG SEGPLKPGAR IFSFDGKDVL RHPTWPQKSV WHGSDPNGRR LTESYCETWR TEAPSATGQA SSLLGGRLLG QSAASCHHAY IVLCIENSFM TASK
Content And Storage -20°C
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.