Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human Endothelin-1 QuantiGlo ELISA Kit
SDP

Catalog No. QET00B Shop All R&D Systems Products
Change view
Click to view available options
Sufficient For:
1 x 96-well Plate
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sufficient For
QET00B 1 x 96-well Plate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. QET00B Supplier R&D Systems™ Supplier No. QET00B
Save 15% on Immunology Workflow Reagents
Use promo code IMMUNO15
View all promotions
Add to Cart
Add to Cart
This item is not returnable. View return policy

Human Endothelin-1 QuantiGlo ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Human Endothelin-1 QuantiGlo ELISA Kit quantifies human Endothelin-1 in cell culture supernates (100 ul), serum (100 ul), edta plasma (100 ul), urine (100 ul).

Specifications

Assay Range 0.34 - 250pg/mL
Assay Sensitivity 0.102pg/mL
Conjugate HRP
Sample Type Cell Culture Supernatant, Plasma (EDTA), Serum, Urine
Specificity Natural and synthetic Human Endothelin-1
Sufficient For 1 x 96-well Plate
For Use With (Application) ELISA
Gene ID (Entrez) 1906
Sample Volume Cell Culture Supernates (100uL),Serum (100uL),EDTA Plasma (100uL),Urine (100uL)
Storage Requirements Store the unopened product at 2° to 8°C. Do not use past expiration date.
Synonym ARCND3, EDN1, endothelin 1, endothelin-1, ET1, HDLCQ7, PPET1, Preproendothelin-1, QME
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.