Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human ENOX1 Full-length ORF (AAH24178.1, 1 a.a. - 643 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89943286 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-943-286 25 μg
89-943-285 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-943-286 Supplier Abnova™ Supplier No. H00055068P01L

Used for AP, Array, ELISA, WB-Re

Electron transport pathways are generally associated with mitochondrial membranes, but non-mitochondrial pathways are also biologically significant. Plasma membrane electron transport pathways are involved in functions as diverse as cellular defense, intracellular redox homeostasis, and control of cell growth and survival. Members of the ecto-NOX family, such as CNOX, or ENOX1, are involved in plasma membrane transport pathways. These enzymes exhibit both a hydroquinone (NADH) oxidase activity and a protein disulfide-thiol interchange activity in series, with each activity cycling every 22 to 26 minutes (Scarlett et al., 2005 [PubMed 15882838]).[supplied by OMIM]

Sequence: MVDAGGVENITQLPQELPQMMAAAADGLGSIAIDTTQLNMSVTDPTAWATAMNNLGMVPVGLPGQQLVSDSICVPGFDPSLNMMTGITPINPMIPGLGLVPPPPPTEVAVVKEIIHCKSCTLFPQNPNLPPPSTRERPPGCKTVFVGGLPENATEEIIQEVFEQCGDITAIRKSKKNFCHIRFAEEFMVDKAIYLSGYRMRLGSSTDKKDSGRLHVDFAQARDDFYEWECKQRMRAREERHRRKLEEDRLRPPSPPAIMHYSEHEAALLAEKLKDDSKFSEAITVLLSWIERGEVNRRSANQFYSMVQSANSHVRRLMNEKATHEQEMEEAKENFKNALTGILTQFEQIVAVFNASTRQKAWDHFSKAQRKNIDIWRKHSEELRNAQSEQLMGIRREEEMEMSDDENCDSPTKKMRVDESALAAQAYALKEENDSLRWQLDAYRNEVELLKQEKEQLFRTEENLTKDQQLQFLQQTMQGMQQQLLTIQEELNNKKSELEQAKEEQSHTQALLKVLQEQLKGTKELVETNGHSHEDSNEINVLTVALVNQDRENNIEKRSQGLKSEKEALLIGIISTFLHVHPFGANIEYLWSYMQQLDSKISANEIEMLLMRLPRMFKQEFTGVGATLEKRWKLCAFEGIKTT

Specifications

Accession Number AAH24178.1
For Use With (Application) Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 55068
Molecular Weight (g/mol) 99.7kDa
Name Human ENOX1 Full-length ORF (AAH24178.1, 1 a.a. - 643 a.a.) Recombinant Protein with GST-tag at N-terminal
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 25 μg
Storage Requirements Store at −80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias CNOX, FLJ10094, PIG38, bA64J21.1, cCNOX
Common Name ENOX1
Gene Symbol ENOX1
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST Tag
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.