Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human ENPP-2/Autotaxin Quantikine ELISA Kit
SDP

Catalog No. DENP20 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DENP20 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. DENP20 Supplier R&D Systems™ Supplier No. DENP20

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Human ENPP-2/Autotaxin Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Human ENPP-2/Autotaxin Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies human ENPP-2/Autotaxin in cell culture supernates (50 ul), serum (10 ul), heparin plasma (10 ul), urine (50 ul), human milk (50 ul).

Specifications

Assay Range 0.78 to 50 ng/mL
Assay Sensitivity 0.157 ng/mL
Conjugate HRP
Product Type Solid Phase Sandwich ELISA
Sample Type Cell Culture Supernatant, Human Milk, Plasma (Heparin), Serum, Urine
Specificity Natural and Recombinant Human ENPP-2
For Use With (Application) ELISA
Gene ID (Entrez) 5168
Gene Symbol ENPP2
Quantity 1 Kit
Sample Volume Cell Culture Supernates (50uL),Serum (10uL),Heparin Plasma (10uL),Urine (50uL),Human Milk (50uL)
Storage Requirements Store the unopened product at 2°C to 8°C. Do not use past expiration date.
Synonym ATX, ATXFLJ26803, ATX-X, AUTOTAXIN, autotaxin-t, EC 3.1.4.39, ectonucleotide pyrophosphatase/phosphodiesterase 2, ectonucleotide pyrophosphatase/phosphodiesterase family member 2, E-NPP 2, ENPP2, Extracellular lysophospholipase D, Lysophosphatidic Acid, LysoPLD, NPP2, PD-IALPHA, PDNP2, PDNP2NPP2, phosphodiesterase I/nucleotide pyrophosphatase 2, plasma lysophospholipase D
Target ENPP-2/Autotaxin
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.