Learn More
Description
- Also known as: B10; NPP3; PDNP3; CD203c; PD-IBETA
- ENPP3 belongs to a family of ectoenzymes that are involved in hydrolysis of extracellular nucleotides.
- These ectoenzymes possess ATPase and ATP pyrophosphatase activities and are type II transmembrane proteins.
- ENPP3 is expressed on activated basophils and mast cells during allergic reactions, cardiac muscle, hepatocytes, and epithelial cells.
- It is upregulated in colon and bile duct carcinomas.
- The insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
- For reasearch use only. Not for use in humans.
Specifications
Specifications
| Gene | ENPP-3/CD203c |
| Accession Number | NP_005012 |
| Concentration | 10 μg at 0.2 μg/μL |
| Formulation | cDNA is provided in 10 mM Tris-Cl, pH 8.5 |
| Insert Type | 2641 bp |
| Gene ID (Entrez) | 5169 |
| Gene Alias | B10, CD203c, CD203c antigen, dJ1005H11.3 (phosphodiesterase I/nucleotide pyrophosphatase 3), dJ914N13.3 (phosphodiesterase I/nucleotide pyrophosphatase 3), ectonucleotide pyrophosphatase/phosphodiesterase 3, ectonucleotide pyrophosphatase/phosphodiesterase family member 3, E-NPP 3, ENPP3, gp130RB13-6, NPP3, PD-IBETA, PDNP3, PDNP3B10, Phosphodiesterase I beta, Phosphodiesterase I/nucleotide pyrophosphatase 3, phosphodiesterase-I beta |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Species | Human |
| Quantity | 10 μg |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
