Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human ENPP1 Full-length ORF (NP_006199.1) Recombinant Protein without tag

Catalog No. 89943290 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-943-290 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-943-290 Supplier Abnova™ Supplier No. H00005167G01
Add to Cart
Add to Cart

Used for AP, Func, Screening

This gene is a member of the ecto-nucleotide pyrophosphatase/phosphodiesterase (ENPP) family. The encoded protein is a type II transmembrane glycoprotein comprising two identical disulfide-bonded subunits. This protein has broad specificity and cleaves a variety of substrates, including phosphodiester bonds of nucleotides and nucleotide sugars and pyrophosphate bonds of nucleotides and nucleotide sugars. This protein may function to hydrolyze nucleoside 5' triphosphates to their corresponding monophosphates and may also hydrolyze diadenosine polyphosphates. Mutations in this gene have been associated with 'idiopathic' infantile arterial calcification, ossification of the posterior longitudinal ligament of the spine (OPLL), and insulin resistance. [provided by RefSeq]

Sequence: MDVGEEPLEKAARARTAKDPNTYKVLSLVLSVCVLTTILGCIFGLKPSCAKEVKSCKGRCFERTFGNCRCDAACVELGNCCLDYQETCIEPEHIWTCNKFRCGEKRLTRSLCACSDDCKDKGDCCINYSSVCQGEKSWVEEPCESINEPQCPAGFETPPTLLFSLDGFRAEYLHTWGGLLPVISKLKKCGTYTKNMRPVYPTKTFPNHYSIVTGLYPESHGIIDNKMYDPKMNASFSLKSKEKFNPEWYKGEPIWVTAKYQGLKSGTFFWPGSDVEINGIFPDIYKMYNGSVPFEERILAVLQWLQLPKDERPHFYTLYLEEPDSSGHSYGPVSSEVIKALQRVDGMVGMLMDGLKELNLHRCLNLILISDHGMEQGSCKKYIYLNKYLGDVKNIKVIYGPAARLRPSDVPDKYYSFNYEGIARNLSCREPNQHFKPYLKHFLPKRLHFAKSDRIEPLTFYLDPQWQLALNPSERKYCGSGFHGSDNVFSNMQALFVGYGPGFKHGIEADTFENIEVYNLMCDLLNLTPAPNNGTHGSLNHLLKNPVYTPKHPKEVHPLVQCPFTRNPRDNLGCSCNPSILPIEDFQTQFNLTVAEEKIIKHETLPYGRPRVLQKENTICLLSQHQFMSGYSQDILMPLWTSYTVDRNDSFSTEDFSNCLYQDFRIPLSPVHKCSFYKNNTKVSYGFLSPPQLNKNSSGIYSEALLTTNIVPMYQSFQVIWRYFHDTLLRKYAEERNGVNVVSGPVFDFDYDGRCDSLENLRQKRRVIRNQEILIPTHFFIVLTSCKDTSQTPLHCENLDTLAFILPHRTDNSESCVHGKHDSSWVEELLMLHRARITDVEHITGLSFYQQRKEPVSDILKLKTHLPTFSQED

Specifications

Accession Number NP_006199.1
For Use With (Application) Antibody Production, Compound Screening, Functional Study
Formulation Liquid
Gene ID (Entrez) 5167
Molecular Weight (g/mol) 99.9kDa
Name ENPP1 (Human) Recombinant Protein
Quantity 10 μg
Immunogen MDVGEEPLEKAARARTAKDPNTYKVLSLVLSVCVLTTILGCIFGLKPSCAKEVKSCKGRCFERTFGNCRCDAACVELGNCCLDYQETCIEPEHIWTCNKFRCGEKRLTRSLCACSDDCKDKGDCCINYSSVCQGEKSWVEEPCESINEPQCPAGFETPPTLLFSLDGFRAEYLHTWGGLLPVISKLKKCGTYTKNMRPVYPTKTFPNHYSIVTGLYPESHGIIDNKMYDPKMNASFSLKSKEKFNPEWYKGEPIWVTAKYQGLKSGTFFWPGSDVEINGIFPDIYKMYNGSVPFEERILAVLQWLQLPKDERPHFYTLYLEEPDSSGHSYGPVSSEVIKALQRVDGMVGMLMDGLKELNLHRCLNLILISDHGMEQGSCKKYIYLNKYLGDVKNIKVIYGPAARLRPSDVPDKYYSFNYEGIARNLSCREPNQHFKPYLKHFLPKRLHFAKSDRIEPLTFYLDPQWQLALNPSERKYCGSGFHGSDNVFSNMQALFVGYGPGFKHGIEADTFENIEVYNLMCDLLNLTPAPNNGTHGSLNHLLKNPVYTPKHPKEVHPLVQCPFTRNPRDNLGCSCNPSILPIEDFQTQFNLTVAEEKIIKHETLPYGRPRVLQKENTICLLSQHQFMSGYSQDILMPLWTSYTVDRNDSFSTEDFSNCLYQDFRIPLSPVHKCSFYKNNTKVSYGFLSPPQLNKNSSGIYSEALLTTNIVPMYQSFQVIWRYFHDTLLRKYAEERNGVNVVSGPVFDFDYDGRCDSLENLRQKRRVIRNQEILIPTHFFIVLTSCKDTSQTPLHCENLDTLAFILPHRTDNSESCVHGKHDSSWVEELLMLHRARITDVEHITGLSFYQQRKEPVSDILKLKTHLPTFSQED
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias M6S1/NPP1/NPPS/PC-1/PCA1/PDNP1
Common Name ENPP1
Gene Symbol ENPP1
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag None
Expression System wheat germ expression system with proprietary liposome technology
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.