Learn More
ACROBiosystems Human ENPP3 2Vials
Supplier:  ACROBiosystems CHEKATP1222VIALS
Encompass_Preferred
The human NPP family contains seven members which can be classified into two groups according to their substrate preferences. The first class comprises the nucleotide-degrading proteins NPP1 3 and 4. NPP3 (CD203c ENPP3) is expressed in multiple organs including on epithelial and mucosal surfaces and notably on basophils and mast cells. Activation of basophils by antigen-bound IgE leads to release of inflammatory mediators and rapid upregulation of NPP3 to the cell surface. This protein is in fact a common marker for diagnosing allergen sensitivity with patient basophils by flow cytometry. Basophils and mast cells mediate the response to certain pathogens as well as acute and chronic allergic reactions. Following activation these cells release ATP which further stimulates them in an autocrine manner. NPP3 upregulation serves to degrade ATP and suppress chronic allergic inflammation (but not the acute response).
Catalog No. 50-253-5274