Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human EphA1 ELISA Kit

Catalog No. EH172RB
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 x 96 Tests
5 x 96 Tests
96 Tests
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EH172RB 96 Tests
EH172RBX5 5 x 96 Tests
EH172RBX10 10 x 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. EH172RB Supplier Invitrogen™ Supplier No. EH172RB
Add to Cart
Add to Cart

ELISA

Human EphA1 quantitates human EphA1 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human EphA1.

EphA1 belongs to the ephrin receptor subfamily of the protein-tyrosine kinase family. EPH and EPH-related receptors have been implicated in mediating developmental events, particularly in the nervous system. Receptors in the EPH subfamily typically have a single kinase domain and an extracellular region containing a Cys-rich domain and 2 fibronectin type III repeats. The ephrin receptors are divided into 2 groups based on the similarity of their extracellular domain sequences and their affinities for binding ephrin-A and ephrin-B ligands. EphA1 is expressed in some human cancer cell lines and has been implicated in carcinogenesis.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number 5730453L17Rik; AL033318; Embryonic stem cell kinase; EPH; Eph receptor A1; EPH tyrosine kinase; EPH tyrosine kinase 1; EPHA1; ephrin receptor EphA1; ephrin type-A receptor 1; EPHT; EPHT1; erythropoietin-producing hepatoma amplified sequence; erythropoietin-producing hepatoma receptor; Esk; hEpha1; mEpha1; MGC163163; oncogene EPH; soluble EPHA1 variant 1; soluble EPHA1 variant 2; Tyrosine-protein kinase receptor EPH; tyrosine-protein kinase receptor ESK
Assay Range 0.085 to 20 ng/mL
Assay Sensitivity 0.085 ng/mL
Conjugate HRP
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 2041
Gene Symbol EPHA1
Interassay CV <12%
Intraassay CV <10
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Quantity 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Storage Requirements 2°C to 8°C
Target EphA1
Test Time 1 hrs 20 mins
Total Assay Time 4 hrs 45 mins
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.