Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human EphB3 ELISA Kit

Catalog No. EH174RBX10
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 x 96 Tests
5 x 96 Tests
96 Tests
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EH174RBX10 10 x 96 Tests
EH174RBX5 5 x 96 Tests
EH174RB 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. EH174RBX10 Supplier Invitrogen™ Supplier No. EH174RBX10
Add to Cart
Add to Cart
Request a Quote

ELISA

Human EphB3 quantitates human EphB3 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human EphB3.

EPHB3 is a receptor tyrosine kinase that belongs to the ephrin receptor family. Members of the Eph family of kinases play important roles in diverse biological processes including nervous system development, angiogenesis and neural synapsis formation and maturation. The ephrin ligands and Eph receptors display reciprocal expression in vivo. Developing and adult neural tissue express nearly all of the Eph receptors and ephrin ligands. Ephs and ephrins play a significant role in angiogenesis.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number AW456895; CAPB; Cek10; Developmental kinase 5; DRT; EK2; ELK; Embryonic kinase 2; EPH receptor B3; EPHB2; Ephb3; EPH-like kinase 2; EPH-like tyrosine kinase 2; EPH-like tyrosine kinase-2; Ephrin B1; Ephrin B2; Ephrin B3; ephrin type-B receptor 3; EphrinB1; EphrinB2; EphrinB3; EPHT3; ERK; Etk2; HEK2; Hek5; HEK-6; human embryo kinase 2; Mdk5; mDK-5; MGC87492; NET; PCBC; Sek4; Tyro5; TYRO6; Tyrosine-protein kinase receptor SEK-4; Tyrosine-protein kinase TYRO6
Assay Range 1.8 to 400 ng/mL
Assay Sensitivity 1.8 ng/mL
Conjugate HRP
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 2049
Gene Symbol EPHB3
Interassay CV <12%
Intraassay CV <10
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Quantity 10 x 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Storage Requirements 2°C to 8°C
Target EphB3
Test Time 1 hrs 20 mins
Total Assay Time 4 hrs 45 mins
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.