Learn More
Sino Biological Human EphB6 MAb 20UL 10197-R031-20
Supplier:  Sino Biological 10197R03120
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human EphB6 / Eph Receptor B6 (rh EphB6 / Eph Receptor B6; Catalog#10197-H08H; NP_004436.1; Met1-Ser579).
Catalog No. 50-241-0065
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.