Learn More
Bioworld Human Epidermal Growth Factor (Recombinant), 100 μg
Supplier:  Bioworld 220603091
Encompass_Preferred
Human Epidermal Growth Factor (Recombinant)Epidermal growth factor has a profound effect on the differentiation of specific cells in vivo and is a potent mitogenic factor for a variety of cultured cells of both ectodermal and mesodermal origin. The EGF precursor is believed to exist as a membrane-bound molecule which is proteolytically cleaved to generate the 53-amino acid peptide hormone that stimulates cells to divide. EGF stimulates the growth of various epidermal and epithelial tissues in vivo and in vitro and of some fibroblasts in cell culture.
Catalog No. 50-253-407