Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Human Epigen Recombinant Protein, PeproTech®

Catalog No. 100515UG
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
100 μg
25 μg
5 μg
500 μg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
100-51-5UG 5 μg
10051100UG 100 μg
100-51-1MG 1 mg
100-51-25UG 25 μg
10051500UG 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 100-51-5UG Supplier Gibco™ Supplier No. 100515UG
Add to Cart
Edge
Add to Cart

Recombinant Protein

Recombinant Human Epigen is a 7.9 kDa monomeric protein, containing 72 amino acid residues, which comprises the EGF-homologous portion of the Epigen precursor. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

Epigen is an EGF-related polypeptide growth factor that signals through the ErbB receptor-1. Promotes the growth of epithelial cells. May stimulate the phosphorylation of EGFR and mitogen-activated protein kinases. It is produced in several tissues, including the testis, liver, and heart, as well as in certain tumor cells. Epigen is mitogenic for fibroblasts and epithelial cells. Human Epigen is initially synthesized as a glycosylated 14.7 kDa transmembrane precursor protein, which is processed by proteolytic cleavage to produce a mature soluble sequence.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q6UW88
For Use With (Application) Functional Assay
Formulation protein with no preservative
Gene ID (Entrez) 255324
Molecular Weight (g/mol) 7.9 kDa
Name Human Epigen
Quantity 5 μg
Source E. coli
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias 2310069M11Rik; ALGV3072; EPG; EPGN; epigen; epithelial mitogen; epithelial mitogen homolog; PRO9904; RGD1560084; UNQ3072/PRO9904
Common Name Epigen
Gene Symbol EPGN
Biological Activity Determined by the dose-dependent stimulation of the proliferation of murine Balb/3T3 cells. The expected ED50 for this effect is 150-300 ng/ml
Conjugate Unconjugated
Recombinant Recombinant
Sequence AVTVTPPITA QQADNIEGPI ALKFSHLCLE DHNSYCINGA CAFHHELEKA ICRCFTGYTG ERCEHLTLTS YA
Content And Storage -20°C
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 98% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.