Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Human EPO (Erythropoietin) Recombinant Protein, PeproTech®

Catalog No. 100641MG
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
10 μg
100 μg
250 μg
50 μg
500 μg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
100-64-1MG 1 mg
10064100UG 100 μg
100-64-10UG 10 μg
10064250UG 250 μg
10064500UG 500 μg
100-64-50UG 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 100-64-1MG Supplier Gibco™ Supplier No. 100641MG
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

PeproTech's Human EPO contains 166 amino acid residues and has a calculated molecular weight of approximately 18.4 kDa. As a result of glycosylation, Recombinant Human EPO migrates with an apparent molecular mass of 37.0 kDa by SDS-PAGE gel, under reducing and non-reducing conditions. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

Erythropoietin (EPO) protein is a growth factor essential for the survival, proliferation, and differentiation of erythroid precursors. EPO is not responsible for early commitment to the lineage, and instead induces the final maturation of partially differentiated erythroid cells and the production of red cell-specific proteins, such as hemoglobin. The EPO receptor (EPOR) is present in higher levels on precursor cells, and expression diminishes with cell maturation.Hepatocytes are the main source of EPO in the fetus, followed by interstitial cells of the kidney after birth and through adulthood. EPO expression is upregulated during hypoxia, and low levels of transcription have also been detected in lung and brain tissue under these conditions. Recent data suggest that EPO may also play a role in the protection of neuronal tissue from damage and initiation of repair.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P01588
For Use With (Application) ELISA, Functional Assay, Western Blot
Formulation protein with no preservative
Gene ID (Entrez) 2056
Molecular Weight (g/mol) 18.4 kDa
Name Human EPO (Erythropoietin)
Quantity 1 mg
Source CHO cells
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias EP; EPO; Epoetin; erythropoietin; MGC138142; MVCD2
Common Name Epo
Gene Symbol EPO
Biological Activity Determined by a cell proliferation assay using TF-1 cells. The expected ED50 for this effect is 0.8-1.0 ng/ml
Conjugate Unconjugated
Recombinant Recombinant
Sequence APPRLICDSR VLERYLLEAK EAENITTGCA EHCSLNENIT VPDTKVNFYA WKRMEVGQQA VEVWQGLALL SEAVLRGQAL LVNSSQPWEP LQLHVDKAVS GLRSLTTLLR ALGAQKEAIS PPDAASAAPL RTITADTFRK LFRVYSNFLR GKLKLYTGEA CRTGDR
Content And Storage -20°C
Expression System CHO cells
Form Lyophilized
Purity or Quality Grade ≥ 90% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.