Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human EPS8 Full-length ORF (NP_004438.3, 1 a.a. - 822 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89943486 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 ug
25 ug
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-943-486 10 ug
89-943-487 25 ug
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-943-486 Supplier Abnova™ Supplier No. H00002059P01S
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

This gene encodes a member of the EPS8 family. This protein contains one PH domain and one SH3 domain. It functions as part of the EGFR pathway, though its exact role has not been determined. Highly similar proteins in other organisms are involved in the transduction of signals from Ras to Rac and growth factor-mediated actin remodeling. Alternate transcriptional splice variants of this gene have been observed but have not been thoroughly characterized. [provided by RefSeq]

Sequence: MNGHISNHPSSFGMYPSQMNGYGSSPTFSQTDREHGSKTSAKALYEQRKNYARDSVSSVSDISQYRVEHLTTFVLDRKDAMITVDDGIRKLKLLDAKGKVWTQDMILQVDDRAVSLIDLESKNELENFPLNTIQHCQAVMHSCSYDSVLALVCKEPTQNKPDLHLFQCDEVKANLISEDIESAISDSKGGKQKRRPDALRMISNADPSIPPPPRAPAPAPPGTVTQVDVRSRVAAWSAWAADQGDFEKPRQYHEQEETPEMMAARIDRDVQILNHILDDIEFFITKLQKAAEAFSELSKRKKNKKGKRKGPGEGVLTLRAKPPPPDEFLDCFQKFKHGFNLLAKLKSHIQNPSAADLVHFLFTPLNMVVQATGGPELASSVLSPLLNKDTIDFLNYTVNGDERQLWMSLGGTWMKARAEWPKEQFIPPYVPRFRNGWEPPMLNFMGATMEQDLYQLAESVANVAEHQRKQEIKRLSTEHSSVSEYHPADGYAFSSNIYTRGSHLDQGEAAVAFKPTSNRHIDRNYEPLKTQPKKYAKSKYDFVARNNSELSVLKDDILEILDDRKQWWKVRNASGDSGFVPNNILDIVRPPESGLGRADPPYTHTIQKQRMEYGPRPADTPPAPSPPPTPAPVPVPLPPSTPAPVPVSKVPANITRQNSSSSDSGGSIVRDSQRHKQLPVDRRKSQMEEVQDELIHRLTIGRSAAQKKFHVPRQNVPVINITYDSTPEDVKTWLQSKGFNPVTVNSLGVLNGAQLFSLNKDELRTVCPEGARVYSQITVQKAALEDSSGSSELQEIMRRRQEKISAAASDSGVESFDEGSSH

Specifications

Accession Number NP_004438.3
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 2059
Molecular Weight (g/mol) 118.3kDa
Name EPS8 (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 ug
Immunogen MNGHISNHPSSFGMYPSQMNGYGSSPTFSQTDREHGSKTSAKALYEQRKNYARDSVSSVSDISQYRVEHLTTFVLDRKDAMITVDDGIRKLKLLDAKGKVWTQDMILQVDDRAVSLIDLESKNELENFPLNTIQHCQAVMHSCSYDSVLALVCKEPTQNKPDLHLFQCDEVKANLISEDIESAISDSKGGKQKRRPDALRMISNADPSIPPPPRAPAPAPPGTVTQVDVRSRVAAWSAWAADQGDFEKPRQYHEQEETPEMMAARIDRDVQILNHILDDIEFFITKLQKAAEAFSELSKRKKNKKGKRKGPGEGVLTLRAKPPPPDEFLDCFQKFKHGFNLLAKLKSHIQNPSAADLVHFLFTPLNMVVQATGGPELASSVLSPLLNKDTIDFLNYTVNGDERQLWMSLGGTWMKARAEWPKEQFIPPYVPRFRNGWEPPMLNFMGATMEQDLYQLAESVANVAEHQRKQEIKRLSTEHSSVSEYHPADGYAFSSNIYTRGSHLDQGEAAVAFKPTSNRHIDRNYEPLKTQPKKYAKSKYDFVARNNSELSVLKDDILEILDDRKQWWKVRNASGDSGFVPNNILDIVRPPESGLGRADPPYTHTIQKQRMEYGPRPADTPPAPSPPPTPAPVPVPLPPSTPAPVPVSKVPANITRQNSSSSDSGGSIVRDSQRHKQLPVDRRKSQMEEVQDELIHRLTIGRSAAQKKFHVPRQNVPVINITYDSTPEDVKTWLQSKGFNPVTVNSLGVLNGAQLFSLNKDELRTVCPEGARVYSQITVQKAALEDSSGSSELQEIMRRRQEKISAAASDSGVESFDEGSSH
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Common Name EPS8
Gene Symbol EPS8
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.