Learn More
Description
- Also known as: Erb; ESRB; ODG8; ESTRB; NR3A2; ER-BETA; ESR-BETA
- ESR2 is a member of the family of estrogen receptors and superfamily of nuclear receptor transcription factors.
- ESR2 contains an N-terminal DNA binding domain and C-terminal ligand binding domain and is localized to the nucleus, cytoplasm, and mitochondria.
- Upon binding to 17beta-estradiol or related ligands, ESR2 forms homo- or hetero-dimers that interact with specific DNA sequences to activate transcription.
- Some isoforms dominantly inhibit the activity of other estrogen receptor family members.
- The insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
- For reasearch use only. Not for use in humans.
Specifications
Specifications
| Gene | hESR2 |
| Accession Number | NP_001428 |
| Concentration | 10 μg at 0.2 μg/μL |
| Formulation | cDNA is provided in 10 mM Tris-Cl, pH 8.5 |
| Insert Type | 1606 bp |
| Gene ID (Entrez) | 2100 |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Species | Human |
| Quantity | 10 μg |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
