Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human ErbB2/Her2 Quantikine ELISA Kit
SDP

Catalog No. DHER20 Shop All Novus Biologicals Products
Change view
Click to view available options
Sufficient For:
1 x 96-well Plate
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sufficient For
DHER20 1 x 96-well Plate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. DHER20 Supplier R&D Systems™ Supplier No. DHER20

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

a 4.5 hour solid-phase ELISA designed to measure human ErbB2 in cell culture supernates, cell lysates, serum, plasma, urine, and human milk.

The Human ErbB2/Her2 Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies human ErbB2/Her2 in cell culture supernates (50 ul), cell lysates (50 ul), serum (10 ul), edta plasma (10 ul), heparin plasma (10 ul), urine (50 ul), human milk (50 ul).

Specifications

Assay Range 78.10 - 5000pg/mL
Assay Sensitivity 14.8pg/mL
Conjugate HRP
Product Line R&D Systems
Sample Type Cell Culture Supernatant, Cell Lysates, Human Milk, Plasma (EDTA, Heparin), Serum, Urine
Specificity Natural and Recombinant Human ErbB2
Sufficient For 1 x 96-well Plate
For Use With (Application) ELISA
Gene ID (Entrez) 2064
Sample Volume Cell Culture Supernates (50uL),Cell Lysates (50uL),Serum (10uL),EDTA Plasma (10uL),Heparin Plasma (10uL),Urine (50uL),Human Milk (50uL)
Storage Requirements Store the unopened product at 2° to 8°C. Do not use past expiration date.
Synonym CD340, CD340 antigen, c-erb B2/neu protein, EC 2.7.10, EGFR2, HER2, HER-2, HER2EC 2.7.10.1, herstatin, Metastatic lymph node gene 19 protein, MLN 19, MLN19, Neu Oncogene, NEUHER-2/neu, neuroblastoma/glioblastoma derived oncogene homolog, NGL, NGLTKR1, p185erbB2, Proto-oncogene c-ErbB-2, Proto-oncogene Neu, receptor tyrosine-protein kinase erbB-2, TKR1, Tyrosine kinase-type cell surface receptor HER2, v-erb-b2 avian erythroblastic leukemia viral oncogene homolog 2(neuro/glioblastoma derived oncogene homolog), v-erb-b2 erythroblastic leukemia viral oncogene homolog 2, neuro/glioblastomaderived oncogene homolog (avian)
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.