Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Human ErbB2/Her2 Recombinant Protein
SDP

Catalog No. NB5965310UG Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
10 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB5965310UG 10 μg
NB5965350UG 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB5965310UG Supplier Novus Biologicals™ Supplier No. NBP25965310UG
Only null left
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results. Applications: SDS-Page

A recombinant protein with a C-Terminal His-tag and corresponding to the amino acids 23-652 of Human ErbB2/Her2 The Recombinant Human ErbB2/Her2 Protein is derived from Baculovirus. The Recombinant Human ErbB2/Her2 Protein has been validated for the following applications: SDS-Page.

Specifications

For Use With (Application) Bioactivity
Formulation In PBS (pH 7.4) containing 10% glycerol
Molecular Weight (g/mol) 70.4 kDa
Name ERBB2 Recombinant Protein
Quantity 10 μg
Source Baculovirus
Storage Requirements Store at 4°C short term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles
Regulatory Status RUO
Endotoxin Concentration <1.0 EU per 1μg of protein (determined by LAL method)
Gene Alias CD340 antigen, CD340
Gene Symbol ERBB2
Product Type Recombinant Protein
Conjugate Unconjugated
Cross Reactivity Human
Recombinant Recombinant
Purity or Quality Grade >85% by SDS-PAGE
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.