Learn More
R&D Systems™ Human ErbB3/Her3 Quantikine ELISA Kit
Specifications
Specifications
| Assay Sensitivity | 15.2 pg/mL |
| Specificity | Natural and Recombinant Human ErbB3 |
| Target Species | Human |
| For Use With (Application) | ELISA |
| Gene ID (Entrez) | 2065 |
| Measuring Range | 62.50 to 4000 pg/mL |
| Quantity | 1 Kit |
| Storage Requirements | Store the unopened product at 2 - 8 °C. Do not use past expiration date. |
| Synonym | c-erbB3, EC 2.7.10, EC 2.7.10.1, ErbB-3, erbB3-S, HER3, HER3c-erbB-3, LCCS2, lethal congenital contracture syndrome 2, MDA-BF-1, MGC88033, p180-ErbB3, p45-sErbB3, p85-sErbB3, Proto-oncogene-like protein c-ErbB-3, receptor tyrosine-protein kinase erbB-3, Tyrosine kinase-type cell surface receptor HER3, v-erb-b2 erythroblastic leukemia viral oncogene homolog 3 (avian) |
| Target | ErbB3/Her3 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.