Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human Erythropoietin Quantikine IVD ELISA Kit
SDP

Catalog No. DEP00B Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DEP00B 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. DEP00B Supplier R&D Systems™ Supplier No. DEP00B

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

Human Erythropoietin Quantikine IVD ELISA Kit

Specifications

Assay Range 2.50 - 200 mU/mL
Assay Sensitivity 0.6 mU/mL
Detection Method Sandwich ELISA
Sample Type Serum, Plasma (EDTA)
Specificity Natural and recombinant human Epo.
Target Species Human
For Use With (Application) ELISA
Gene ID (Entrez) 2056
Quantity 1 Kit
Storage Requirements Store the unopened product at 2 - 8 °C. Do not use past expiration date.
Synonym ECYT5, EP, EPO, epoetin, erythropoietin, MGC138142, MVCD2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.