Learn More
Description
Includes
- 1.NB100-1831: SSEA-1 Antibody (MC-480)
- 2.NB100-1832: SSEA-3 Antibody (MC-631)
- 3.NB100-1833: TRA-1-81 Antibody (TRA-1-81)
- 4.NB100-2379: OCT4 Antibody
- 5.NB100-588: Nanog Antibody
- 6.NB100-730: TRA-1-60 (R) Antibody (TRA-1-60(R))
Specifications
Specifications
| Antigen | Human ESC Marker |
| Applications | Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot |
| Conjugate | Unconjugated |
| Dilution | Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin |
| Quantity | 6 Vials |
| Research Discipline | Embryonic Stem Cell Markers, Stem Cell Markers |
| Test Specificity | NB100-588 is specific for mouse Nanog protein. NB110-56937:A synthetic peptide corresponding to residues in the 5th cadherin domain of human E-cadherin was used. NB100-1832 Rat Monoclonal to SSEA3 (MC-631) - Embryonic Stem Cell Marker NB100-730 recognizes antigens that are associated with a pericellular matrix proteoglycan. TRA-1-60 reacts with a sialidase-sensitive epitope. NB100-1833 is specific to TRA-1-81 - Embryonic Stem Cell Marker |
| Includes | 1.NB100-1831: SSEA-1 Antibody (MC-480) 2.NB100-1832: SSEA-3 Antibody (MC-631) 3.NB100-1833: TRA-1-81 Antibody (TRA-1-81) 4.NB100-2379: OCT4 Antibody 5.NB100-588: Nanog Antibody 6.NB100-730: TRA-1-60 (R) Antibody (TRA-1-60(R)) |
| Target Species | Human, Mouse |
| Content And Storage | Storage is content dependent. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
