Learn More
Abcam Human ESM1 ELISA Kit (Endocan), 1 x 96 Tests
Supplier:  Abcam AB2137761X96TESTS
Encompass_Preferred
Human ESM1 ELISA Kit (Endocan) is a Sandwich (quantitative) ELISA kit for the measurement of Human ESM1 (Endocan) in Human in Cell Lysate, Cell culture supernatant, EDTA Plasma, Heparin Plasma, Serum samples.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC2602509
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.