Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human F-box protein 15/FBXO15 Alexa Fluor 488-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. IC6035G100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
IC6035G100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. IC6035G100 Supplier R&D Systems Supplier No. IC6035G100UG
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

F-box protein 15/FBXO15 Monoclonal specifically detects F-box protein 15/FBXO15 in Human samples. It is validated for Intracellular Staining by Flow Cytometry.

Specifications

Antigen F-box protein 15/FBXO15
Applications Flow Cytometry
Classification Monoclonal
Clone 612834
Concentration 0.2 mg/mL
Conjugate Alexa Fluor 488
Dilution Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
Gene Accession No. Q8NCQ5-2
Gene Alias F-box only protein 15, Fbox protein 15, F-box protein 15, FBX15, FBX15MGC39671, FBXO15
Gene Symbols FBXO15
Host Species Mouse
Immunogen E. coli-derived recombinant human F-box protein 15/FBXO15 isoform 2 Asn298-Tyr434 Accession # Q8NCQ5-2
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 201456
Test Specificity Detects human F-box protein 15/FBXO15 in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant mouse F-box protein 15 is observed.
Target Species Human
Content And Storage Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Isotype IgG2a
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.