Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human FAAH2 Full-length ORF (NP_777572.1, 1 a.a. - 532 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89933286 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
2 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-933-286 2 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-933-286 Supplier Abnova™ Supplier No. H00158584P01
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

This gene encodes a fatty acid amide hydrolase that shares a conserved protein motif with the amidase signature family of enzymes. The encoded enzyme is able to catalyze the hydrolysis of a broad range of bioactive lipids, including those from the three main classes of fatty acid amides; N-acylethanolamines, fatty acid primary amides and N-acyl amino acids. This enzyme has a preference for monounsaturated acyl chains as a substrate

Sequence: MAPSFTARIQLFLLRALGFLIGLVGRAALVLGGPKFASKTPRPVTEPLLLLSGMQLAKLIRQRKVKCIDVVQAYINRIKDVNPMINGIVKYRFEEAMKEAHAVDQKLAEKQEDEATLENKWPFLGVPLTVKEAFQLQGMPNSSGLMNRRDAIAKTDATVVALLKGAGAIPLGITNCSELCMWYESSNKIYGRSNNPYDLQHIVGGSSGGEGCTLAAACSVIGVGSDIGGSIRMPAFFNGIFGHKPSPGVVPNKGQFPLAVGAQELFLCTGPMCRYAEDLAPMLKVMAGPGIKRLKLDTKVHLKDLKFYWMEHDGGSFLMSKVDQDLIMTQKKVVVHLETILGASVQHVKLKKMKYSFQLWIAMMSAKGHDGKEPVKFVDLLGDHGKHVSPLWELIKWCLGLSVYTIPSIGLALLEEKLRYSNEKYQKFKAVEESLRKELVDMLGDDGVFLYPSHPTVAPKHHVPLTRPFNFAYTGVFSALGLPVTQCPLGLNAKGLPLGIQVVAGPFNDHLTLAVAQYLEKTFGGWVCPGKF

Specifications

Accession Number NP_777572.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 158584
Molecular Weight (g/mol) 84.7kDa
Name FAAH2 (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 2 μg
Immunogen MAPSFTARIQLFLLRALGFLIGLVGRAALVLGGPKFASKTPRPVTEPLLLLSGMQLAKLIRQRKVKCIDVVQAYINRIKDVNPMINGIVKYRFEEAMKEAHAVDQKLAEKQEDEATLENKWPFLGVPLTVKEAFQLQGMPNSSGLMNRRDAIAKTDATVVALLKGAGAIPLGITNCSELCMWYESSNKIYGRSNNPYDLQHIVGGSSGGEGCTLAAACSVIGVGSDIGGSIRMPAFFNGIFGHKPSPGVVPNKGQFPLAVGAQELFLCTGPMCRYAEDLAPMLKVMAGPGIKRLKLDTKVHLKDLKFYWMEHDGGSFLMSKVDQDLIMTQKKVVVHLETILGASVQHVKLKKMKYSFQLWIAMMSAKGHDGKEPVKFVDLLGDHGKHVSPLWELIKWCLGLSVYTIPSIGLALLEEKLRYSNEKYQKFKAVEESLRKELVDMLGDDGVFLYPSHPTVAPKHHVPLTRPFNFAYTGVFSALGLPVTQCPLGLNAKGLPLGIQVVAGPFNDHLTLAVAQYLEKTFGGWVCPGKF
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias AMDD/FAAH-2/FLJ31204/RP11-479E16
Common Name FAAH2
Gene Symbol FAAH2
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.