Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Human FABP2/I-FABP Quantikine ELISA Kit
SDP

Catalog No. DFBP20 Shop All Novus Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DFBP20 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. DFBP20 Supplier R&D Systems™ Supplier No. DFBP20
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Human FABP2/I-FABP Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

The Human FABP2/I-FABP Quantikine ELISA Kit quantifies human FABP2/I-FABP in cell culture supernates (50 ul), cell lysates (50 ul), serum (10 ul), edta plasma (10 ul), heparin plasma (10 ul), urine (50 ul).

Specifications

Assay Range 15.60 - 1000pg/mL
Assay Sensitivity 6.21pg/mL
Detectable Analytes FABP2/I-FABP
Assay Solid Phase ELISA
Conjugate HRP
Cross Reactivity <0.5% cross-reactivity observed with available related molecules; <50% cross-species reactivity observed with species tested
Immunoassay Kit Format 96-well Strip Plate
Product Line R&D Systems
Product Type ELISA Kit
Sample Type Cell Culture Supernatant, Cell Lysates, Plasma (EDTA, Heparin), Serum, Urine
Specificity Natural and Recombinant Human FABP2
Target Species Human
For Use With (Application) ELISA
Gene ID (Entrez) 2169, 25598
Host Species Human
Quantity 1 Kit
Sample Volume Cell Culture Supernates (50uL),Cell Lysates (50uL),Serum (10uL),EDTA Plasma (10uL),Heparin Plasma (10uL),Urine (50uL)
Storage Requirements Store the unopened product at 2° to 8°C. Do not use past expiration date.
Synonym FABPI, fatty acid binding protein 2, intestinal, Fatty acid-binding protein 2, fatty acid-binding protein, intestinal, IFABP, I-FABP, I-FABPMGC133132, Intestinal FABP, Intestinal-type fatty acid-binding protein
Total Assay Time 4.5 hr.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.