Learn More
Human FABP7/B-FABP Antibody, R&D Systems™



Description
Specifications
Specifications
| Antigen | FABP7/B-FABP |
| Applications | Immunocytochemistry, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Simple Western, Western Blot |
| Classification | Polyclonal |
| Concentration | LYOPH |
| Conjugate | Unconjugated |
| Dilution | Western Blot 0.1 ug/mL, Simple Western 50 ug/mL, Immunohistochemistry 5-15 ug/mL |
| Gene Accession No. | O15540 |
| Gene Alias | BFABP, B-FABP, B-FABPDKFZp547J2313, BLBP, BLBPMRG, Brain lipid-binding protein, Brain-type fatty acid-binding protein, FABPBbrain lipid binding protein, fatty acid binding protein 7, brain, Fatty acid-binding protein 7, fatty acid-binding protein, brain, Mammary-derived growth inhibitor related, mammary-derived growth inhibitor-related |
| Gene Symbols | FABP7 |
| Host Species | Goat |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.