Learn More
Haematologic Technologies Inc HUMAN FACTOR IXA 100UG
Supplier:  Haematologic Technologies Inc HCIXA0050
Encompass_Preferred
Coagulation protein, Human factor IXa, 100 micrograms, Native enzyme, Mode of Action: Enzyme component of the Factor Xase complex, Research use only, 50% (v/v) glycerol/water, store at -20 Celsius.
Catalog No. 50-883-392
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.