Learn More
Haematologic Technologies Inc HUMAN FACTOR XA 100UG
Supplier:  Haematologic Technologies Inc HCXA0060
Encompass_Preferred
Coagulation protein, Human factor Xa, 100 micrograms, Native enzyme, Mode of Action: Enzyme component of the prothrombinase complex, Research use only, 50% (v/v) glycerol/water, store at -20 Celsius.
Catalog No. 50-883-452