Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human FAM59B Synthetic Peptide

Catalog No. PEP0960
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PEP0960 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PEP0960 Supplier Invitrogen™ Supplier No. PEP0960
Add to Cart
Add to Cart

Synthetic Peptide

FAM59B Synthetic Peptide, PEP-0960, from Invitrogen.

FAM59B is related to the FAM59A protein (also known as GAREM). While little is known of the role of the FAM59B protein, FAM59A is a downstream molecule in the EGF signaling pathway and binds Grb2, an adaptor protein crucial to the transduction of growth signals from the plasma membrane to the nucleus. Like FAM59A, FAM59B may play a role in ligand-mediated signaling pathway of the EGF receptor and the tumorigenesis of cells.
TRUSTED_SUSTAINABILITY

Specifications

Gene ID (Entrez) 150946
Species Human
Purification Method purified
Accession Number Q75VX8
Common Name Human FAM59B
Concentration 200 μg/mL
Content And Storage -20°C
Description Synthetic Peptide
Format Liquid
Formulation PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2
For Use With (Application) Blocking Assay, Control
Gene Symbol GAREM2
Lyophilized No
Preservative 0.02% sodium azide
Product Type Peptide
Quantity 50 μg
Regulatory Status RUO - research use only
Storage Buffer PBS
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.