Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human FANCC Full-length ORF (NP_000127.2, 1 a.a. - 558 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89944050 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
10 ug
25 ug
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-944-050 10 ug
89-944-051 25 ug
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 89-944-050 Supplier Abnova™ Supplier No. H00002176P01S
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

The Fanconi anemia complementation group (FANC) currently includes FANCA, FANCB, FANCC, FANCD1 (also called BRCA2), FANCD2, FANCE, FANCF, FANCG, FANCI, FANCJ (also called BRIP1), FANCL, FANCM and FANCN (also called PALB2). The previously defined group FANCH is the same as FANCA. Fanconi anemia is a genetically heterogeneous recessive disorder characterized by cytogenetic instability, hypersensitivity to DNA crosslinking agents, increased chromosomal breakage, and defective DNA repair. The members of the Fanconi anemia complementation group do not share sequence similarity; they are related by their assembly into a common nuclear protein complex. This gene encodes the protein for complementation group C. [provided by RefSeq]

Sequence: MAQDSVDLSCDYQFWMQKLSVWDQASTLETQQDTCLHVAQFQEFLRKMYEALKEMDSNTVIERFPTIGQLLAKACWNPFILAYDESQKILIWCLCCLINKEPQNSGQSKLNSWIQGVLSHILSALRFDKEVALFTQGLGYAPIDYYPGLLKNMVLSLASELRENHLNGFNTQRRMAPERVASLSRVCVPLITLTDVDPLVEALLICHGREPQEILQPEFFEAVNEAILLKKISLPMSAVVCLWLRHLPSLEKAMLHLFEKLISSERNCLRRIECFIKDSSLPQAACHPAIFRVVDEMFRCALLETDGALEIIATIQVFTQCFVEALEKASKQLRFALKTYFPYTSPSLAMVLLQDPQDIPRGHWLQTLKHISELLREAVEDQTHGSCGGPFESWFLFIHFGGWAEMVAEQLLMSAAEPPTALLWLLAFYYGPRDGRQQRAQTMVQVKAVLGHLLAMSRSSSLSAQDLQTVAGQGTDTDLRAPAQQLIRHLLLNFLLWAPGGHTIAWDVITLMAHTAEITHEIIGFLDQTLYRWNRLGIESPRSEKLARELLKELRTQV

Specifications

Accession Number NP_000127.2
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 2176
Molecular Weight (g/mol) 89.8kDa
Name FANCC (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 10 ug
Immunogen MAQDSVDLSCDYQFWMQKLSVWDQASTLETQQDTCLHVAQFQEFLRKMYEALKEMDSNTVIERFPTIGQLLAKACWNPFILAYDESQKILIWCLCCLINKEPQNSGQSKLNSWIQGVLSHILSALRFDKEVALFTQGLGYAPIDYYPGLLKNMVLSLASELRENHLNGFNTQRRMAPERVASLSRVCVPLITLTDVDPLVEALLICHGREPQEILQPEFFEAVNEAILLKKISLPMSAVVCLWLRHLPSLEKAMLHLFEKLISSERNCLRRIECFIKDSSLPQAACHPAIFRVVDEMFRCALLETDGALEIIATIQVFTQCFVEALEKASKQLRFALKTYFPYTSPSLAMVLLQDPQDIPRGHWLQTLKHISELLREAVEDQTHGSCGGPFESWFLFIHFGGWAEMVAEQLLMSAAEPPTALLWLLAFYYGPRDGRQQRAQTMVQVKAVLGHLLAMSRSSSLSAQDLQTVAGQGTDTDLRAPAQQLIRHLLLNFLLWAPGGHTIAWDVITLMAHTAEITHEIIGFLDQTLYRWNRLGIESPRSEKLARELLKELRTQV
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias FA3/FAC/FACC/FLJ14675
Common Name FANCC
Gene Symbol FANCC
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.