Learn More
ACROBiosystems Human FAP 10mg
Supplier:  ACROBiosystems MBSK07710MG
Encompass_Preferred
The biotinylated FAP protein was conjugated to streptavidin magnetic beads. This pre-coupled magnetic bead product can capture the anti-FAP antibody from various assay systems. The beads are in uniform size narrow size distribution with large surface area and unique surface coating which can help you get the best performance and highly reproducible results. This FAP coupled magnetic beads will bring great convenience with minimum non-specific binding and developed protocols. This ready-to-use product could greatly save your time and hassle.
Catalog No. 50-253-4452
Only null left
Add to cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.