Learn More
Sino Biological Human Fas Ligand / FASLG / CD95L Protein (His Tag) 50ug
Supplier:  Sino Biological 10244H07Y50
Encompass_Preferred
A DNA sequence encoding human FASLG (NP_000630.1) (Pro134-Leu281) was expressed with a polyhistidine tag at the N-terminus.
Catalog No. 50-224-7073
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.