Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human FASTK Full-length ORF (NP_006703.1, 1 a.a. - 549 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89944093 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
2 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-944-093 2 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-944-093 Supplier Abnova™ Supplier No. H00010922P01
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

The protein encoded by this gene is a member of the serine/threonine protein kinase family. This kinase was shown to be activated rapidly during Fas-mediated apoptosis in Jurkat cells. In response to Fas receptor ligation, it phosphorylates TIA1, an apoptosis-promoting nuclear RNA-binding protein. The encoded protein is a strong inducer of lymphocyte apoptosis. Two transcript variants encoding different isoforms have been found for this gene. Other variants exist, but their full-length natures have not yet been determined. [provided by RefSeq]

Sequence: MRRPRGEPGPRAPRPTEGATCAGPGESWSPSPNSMLRVLLSAQTSPARLSGLLLIPPVQPCCLGPSKWGDRPVGGGPSAGPVQGLQRLLEQAKSPGELLRWLGQNPSKVRAHHYSVALRRLGQLLGSRPRPPPVEQVTLQDLSQLIIRNCPSFDIHTIHVCLHLAVLLGFPSDGPLVCALEQERRLRLPPKPPPPLQPLLRGGQGLEAALSCPRFLRYPRQHLISSLAEARPEELTPHVMVLLAQHLARHRLREPQLLEAIAHFLVVQETQLSSKVVQKLVLPFGRLNYLPLEQQFMPCLERILAREAGVAPLATVNILMSLCQLRCLPFRALHFVFSPGFINYISGTPHALIVRRYLSLLDTAVELELPGYRGPRLPRRQQVPIFPQPLITDRARCKYSHKDIVAEGLRQLLGEEKYRQDLTVPPGYCTDFLLCASSSGAVLPVRTQDPFLPYPPRSCPQGQAASSATTRDPAQRVVLVLRERWHFCRDGRVLLGSRALRERHLGLMGYQLLPLPFEELESQRGLPQLKSYLRQKLQALGLRWGPEGG

Specifications

Accession Number NP_006703.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 10922
Molecular Weight (g/mol) 87.5kDa
Name FASTK (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 2 μg
Immunogen MRRPRGEPGPRAPRPTEGATCAGPGESWSPSPNSMLRVLLSAQTSPARLSGLLLIPPVQPCCLGPSKWGDRPVGGGPSAGPVQGLQRLLEQAKSPGELLRWLGQNPSKVRAHHYSVALRRLGQLLGSRPRPPPVEQVTLQDLSQLIIRNCPSFDIHTIHVCLHLAVLLGFPSDGPLVCALEQERRLRLPPKPPPPLQPLLRGGQGLEAALSCPRFLRYPRQHLISSLAEARPEELTPHVMVLLAQHLARHRLREPQLLEAIAHFLVVQETQLSSKVVQKLVLPFGRLNYLPLEQQFMPCLERILAREAGVAPLATVNILMSLCQLRCLPFRALHFVFSPGFINYISGTPHALIVRRYLSLLDTAVELELPGYRGPRLPRRQQVPIFPQPLITDRARCKYSHKDIVAEGLRQLLGEEKYRQDLTVPPGYCTDFLLCASSSGAVLPVRTQDPFLPYPPRSCPQGQAASSATTRDPAQRVVLVLRERWHFCRDGRVLLGSRALRERHLGLMGYQLLPLPFEELESQRGLPQLKSYLRQKLQALGLRWGPEGG
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias FAST
Common Name FASTK
Gene Symbol FASTK
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.