Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human FBXL3 Full-length ORF (NP_036290.1, 1 a.a. - 428 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89944150 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
2 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-944-150 2 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-944-150 Supplier Abnova™ Supplier No. H00026224P01
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

This gene encodes a member of the F-box protein family which is characterized by an approximately 40 amino acid motif, the F-box. The F-box proteins constitute one of the four subunits of ubiquitin protein ligase complex called SCFs (SKP1-cullin-F-box), which function in phosphorylation-dependent ubiquitination. The F-box proteins are divided into 3 classes: Fbws containing WD-40 domains, Fbls containing leucine-rich repeats, and Fbxs containing either different protein-protein interaction modules or no recognizable motifs. The protein encoded by this gene belongs to the Fbls class and, in addition to an F-box, contains several tandem leucine-rich repeats and is localized in the nucleus. [provided by RefSeq]

Sequence: MKRGGRDSDRNSSEEGTAEKSKKLRTTNEHSQTCDWGNLLQDIILQVFKYLPLLDRAHASQVCRNWNQVFHMPDLWRCFEFELNQPATSYLKATHPELIKQIIKRHSNHLQYVSFKVDSSKESAEAACDILSQLVNCSLKTLGLISTARPSFMDLPKSHFISALTVVFVNSKSLSSLKIDDTPVDDPSLKVLVANNSDTLKLLKMSSCPHVSPAGILCVADQCHGLRELALNYHLLSDELLLALSSEKHVRLEHLRIDVVSENPGQTHFHTIQKSSWDAFIRHSPKVNLVMYFFLYEEEFDPFFRYEIPATHLYFGRSVSKDVLGRVGMTCPRLVELVVCANGLRPLDEELIRIAERCKNLSAIGLGECEVSCSAFVEFVKMCGGRLSQLSIMEEVLIPDQKYSLEQIHWEVSKHLGRVWFPDMMPTW

Specifications

Accession Number NP_036290.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 26224
Molecular Weight (g/mol) 75.1kDa
Name FBXL3 (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 2 μg
Immunogen MKRGGRDSDRNSSEEGTAEKSKKLRTTNEHSQTCDWGNLLQDIILQVFKYLPLLDRAHASQVCRNWNQVFHMPDLWRCFEFELNQPATSYLKATHPELIKQIIKRHSNHLQYVSFKVDSSKESAEAACDILSQLVNCSLKTLGLISTARPSFMDLPKSHFISALTVVFVNSKSLSSLKIDDTPVDDPSLKVLVANNSDTLKLLKMSSCPHVSPAGILCVADQCHGLRELALNYHLLSDELLLALSSEKHVRLEHLRIDVVSENPGQTHFHTIQKSSWDAFIRHSPKVNLVMYFFLYEEEFDPFFRYEIPATHLYFGRSVSKDVLGRVGMTCPRLVELVVCANGLRPLDEELIRIAERCKNLSAIGLGECEVSCSAFVEFVKMCGGRLSQLSIMEEVLIPDQKYSLEQIHWEVSKHLGRVWFPDMMPTW
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias FBL3/FBL3A/FBXL3A
Common Name FBXL3
Gene Symbol FBXL3
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.