Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human FBXO33 Full-length ORF (NP_976046.1, 1 a.a. - 555 a.a.) Recombinant Protein with GST-tag at N-terminal

Catalog No. 89944217 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
2 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-944-217 2 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-944-217 Supplier Abnova™ Supplier No. H00254170P01
Add to Cart
Add to Cart

Used for AP, Array, ELISA, WB-Re

Members of the F-box protein family, such as FBXO33, are characterized by an approximately 40-amino acid F-box motif. SCF complexes, formed by SKP1 (MIM 601434), cullin (see CUL1; MIM 603134), and F-box proteins, act as protein-ubiquitin ligases. F-box proteins interact with SKP1 through the F box, and they interact with ubiquitination targets through other protein interaction domains (Jin et al., 2004 [PubMed 15520277]).[supplied by OMIM]

Sequence: MLLFLSVPQPRPPGARTRAGAARVARWRRLRLQQLRRLRGLLRVLRGRPGAGSRRRGRMALCGQAAGAASLPSELIVHIFSFLPAPDRLRASASCSHWRECLFYPALWPQLRICLRVSPAEQPRLEFLMRKCGWFVRELRVEFAAENYLSGGGPGDGGGADTGTGGEEVEALQLSARWLEVLRTYLELVLCVLVSIRNNRNLQKFSLFGDISVLQQQGSLSNTYLSKVDPDGKKIKQIQQLFEEILSNSRQLKWLSCGFMLEIVTPTSLSSLSNAVANTMEHLSLLDNNIPGNSTLITAVELERFVNLHSLALDFCDFTAEMARVLTDSNHVPLQRLSLLVHNVSVMHKSLDNMPNDEHWKALSRKSTSFRVYIMAFDIKSEDMLKILKPSIPLERIHFDSYITCVSGAIVDLISRQYDKFLTHFILMNDVIDTSGFPDLSDNRNEDPLVLLAWRCTKLSLLAIHGYTVWAHNLIAIARLRGSDLKVLEVTEESIDFDQGELADQDVDPVHNLIEQVSLGLGQPWHAVMDIESLSVFTEPNRHFYREMQSFSEDI

Specifications

Accession Number NP_976046.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 254170
Molecular Weight (g/mol) 89kDa
Name FBXO33 (Human) Recombinant Protein (P01)
Purification Method Glutathione Sepharose 4 Fast Flow
Quality Control Testing 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantity 2 μg
Immunogen MLLFLSVPQPRPPGARTRAGAARVARWRRLRLQQLRRLRGLLRVLRGRPGAGSRRRGRMALCGQAAGAASLPSELIVHIFSFLPAPDRLRASASCSHWRECLFYPALWPQLRICLRVSPAEQPRLEFLMRKCGWFVRELRVEFAAENYLSGGGPGDGGGADTGTGGEEVEALQLSARWLEVLRTYLELVLCVLVSIRNNRNLQKFSLFGDISVLQQQGSLSNTYLSKVDPDGKKIKQIQQLFEEILSNSRQLKWLSCGFMLEIVTPTSLSSLSNAVANTMEHLSLLDNNIPGNSTLITAVELERFVNLHSLALDFCDFTAEMARVLTDSNHVPLQRLSLLVHNVSVMHKSLDNMPNDEHWKALSRKSTSFRVYIMAFDIKSEDMLKILKPSIPLERIHFDSYITCVSGAIVDLISRQYDKFLTHFILMNDVIDTSGFPDLSDNRNEDPLVLLAWRCTKLSLLAIHGYTVWAHNLIAIARLRGSDLKVLEVTEESIDFDQGELADQDVDPVHNLIEQVSLGLGQPWHAVMDIESLSVFTEPNRHFYREMQSFSEDI
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Gene Alias Fbx33/c14_5247
Common Name FBXO33
Gene Symbol FBXO33
Species Wheat Germ (in vitro)
Recombinant Recombinant
Protein Tag GST
Expression System wheat germ expression system
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.