Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human Fc gamma RIII (CD16) Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. p-7163437
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB4325SP 25 μg
MAB4325100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB4325SP Supplier R&D Systems Supplier No. MAB4325SP
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Fc gamma RIII (CD16) Monoclonal specifically detects Fc gamma RIII (CD16) in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, CyTOF-ready.

Specifications

Antigen Fc gamma RIII (CD16)
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, CyTOF
Classification Monoclonal
Clone 1001049
Conjugate Unconjugated
Dilution Western Blot 2 μg/mL, Flow Cytometry 0.25 μg/106 cells, Immunohistochemistry 5-25 μg/mL, Immunocytochemistry 8-25 μg/mL, CyTOF-ready
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
Gene Accession No. P08637
Gene Alias CD16, CD16A, CD16b, Fc fragment of IgG receptor IIIa, FCG3, FCGR3, FCGR3A, FCGR3B, FCGRIII, FcgRIIIA, FcgRIIIB, FcR-10, FCRIII, FCRIIIA, FCRIIIB, IGFR3, IMD20
Host Species Mouse
Immunogen Human embryonic kidney cell, HEK293-derived human Fc gamma RIIIA/CD16a, Gly17-Gln208, Accession # P08637
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Primary or Secondary Primary
Gene ID (Entrez) 2214
Test Specificity Detects human Fc gamma RIIIA/CD16a in direct ELISAs. In direct ELISAs, 60% cross-reactivity with Fc gamma RIIIB/CD16b is observed.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution.
Form Lyophilized
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.