Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Human FCAR (P24071, Gln 22 - Asn 227) Partial Recombinant Protein with C-terminal 6His tag expressed in HEK293 cells

Catalog No. 89944298 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
20 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-944-298 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-944-298 Supplier Abnova™ Supplier No. P3686
Add to Cart
Add to Cart

Used for SDS-PAGE

This gene is a member of the immunoglobulin gene superfamily and encodes a receptor for the Fc region of IgA. The receptor is a transmembrane glycoprotein present on the surface of myeloid lineage cells such as neutrophils, monocytes, macrophages, and eosinophils, where it mediates immunologic responses to pathogens. It interacts with IgA-opsonized targets and triggers several immunologic defense processes, including phagocytosis, antibody-dependent cell-mediated cytotoxicity, and stimulation of the release of inflammatory mediators. Multiple alternatively spliced transcript variants encoding different isoforms have been described for this gene. [provided by RefSeq]

Specifications

Accession Number P24071
For Use With (Application) SDS-PAGE
Formulation Lyophilized
Gene ID (Entrez) 2204
Molecular Weight (g/mol) 45-50kDa
Name FCAR (Human) Recombinant Protein
Preparation Method Mammalian cell (HEK293) expression system
Purification Method Ion exchange column and HPLC reverse phase column
Quantity 20 μg
Storage Requirements Store at -20°C on dry atmosphere. After reconstitution with deionized water, store at 4°C for one month. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing.
Regulatory Status RUO
Endotoxin Concentration <0.1ng/μg (1 EU/μg)
Gene Alias CD89
Common Name FCAR
Gene Symbol FCAR
Species Human
Recombinant Recombinant
Protein Tag His
Expression System Mammalian cell (HEK293) expression system
Form Lyophilized
Purity or Quality Grade >90% by SDS-PAGE and HPLC
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.