Learn More
Sino Biological Human CD16a / FCGR3A Protein (176 Val) (His & AVI Tag) 1mg
Supplier:  Sino Biological 10389H27H11
Encompass_Preferred
A DNA sequence encoding the extracellular domain (Met 1-Gln 208) of human CD16a (AAH17865.1) was fused with a c-terminal polyhistidine tagged AVI tag at the C-terminus.
Catalog No. 50-255-5336
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.