Learn More
Description
- Also known as: FcRY; FCRL2; FCRLY; FREB2; FCRLM2; FREB-2
- FCRLB belongs to the Fc receptor family. Fc receptors are involved in phagocytosis, antibody-dependent cell cytotoxicity, immediate hypersensitivity, and transcytosis of immunoglobulins via their ability to bind immunoglobulin (Ig) constant regions
- FCRLB is expressed at a low level in placenta and in B lineage cells of the germinal center
- It is upregulated in B cells by BAFF and LPS
- Among non-hematopoietic cells, FCRLB is expressed in fibroblasts, melanocytes, and melanoma
- It is preferentially expressed in nonproliferating cells and at the onset of apoptosis
- The FCRLB/FCRY insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided
- For reasearch use only. Not for use in humans.
Specifications
Specifications
| Gene | hFCRLB |
| Accession Number | NP_001002901 |
| Concentration | 10 μg at 0.2 μg/μL |
| Formulation | cDNA is provided in 10 mM Tris-Cl, pH 8.5 |
| Insert Type | 1294 bp |
| Gene ID (Entrez) | 127943 |
| Content And Storage | Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Species | Human |
| Quantity | 10 μg |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
