Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Human FDA Standard Tissue MicroArray (Normal)
SDP

Catalog No. NBP278123 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
5 Slides
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP278123 5 Slides
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NBP278123 Supplier Novus Biologicals™ Supplier No. NBP278123
Add to Cart
Add to Cart
This item is not returnable. View return policy

The panel contains 5 to 8 different tissues on one slide.

Specifications

Quantity 5 Slides
Species Human
For Use With (Application) Please bake slides at 60C for 30 minutes before use.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.