Description
- It includes a dissected left ovary that displays the maturation process of the ovum
- Represents ovulation, fertilization, blastocyte formation and implantation
- Structures are numbered and identifiable on the included key
- Models are mounted on plastic bases and made exclusively from high quality medical PVC
Specifications
Specifications
| Material | PVC |
| Gender | Female |
| Product Type | Internal Reproductive Organ Model (Female) |
| Grade Level | High School |