Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Human FGF-10, Animal-Free Recombinant Protein, PeproTech®

Catalog No. AF10026100
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
100 μg
25 μg
500 μg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF10026100 100 μg
AF100261MG 1 mg
AF1002625UG 25 μg
AF10026500 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. AF10026100 Supplier Gibco™ Supplier No. AF10026100UG
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

Recombinant Human FGF-10 is a 19.3 kDa protein consisting of 170 amino acid residues. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

FGF10 is a member of the fibroblast growth factor (FGF) family. FGF family members possess broad mitogenic and cell survival activities, and are involved in a variety of biological processes, including embryonic development, cell growth, morphogenesis, tissue repair, tumor growth and invasion. This protein exhibits mitogenic activity for keratinizing epidermal cells, but essentially no activity for fibroblasts, which is similar to the biological activity of FGF7. Studies of the mouse homolog of suggested that this gene is required for embryonic epidermal morphogenesis including brain development, lung morphogenesis, and initiation of lim bud formation. This protein is also implicated to be a primary factor in the process of wound healing.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number O15520
For Use With (Application) Functional Assay
Formulation protein with no preservative
Gene ID (Entrez) 2255
Molecular Weight (g/mol) 19.3 kDa
Name Human FGF-10, Animal-Free
Quantity 100 μg
Source E. coli
Regulatory Status RUO
Endotoxin Concentration <0.1 EU/ μg
Gene Alias AEY17; BB213776; FGF; Fgf10; FGF-10; Fibroblast growth factor; Fibroblast growth factor 10; Gsfaey17; Keratinocyte growth factor 2; produced by fibroblasts of urinary bladder lamina propria
Common Name FGF10
Gene Symbol FGF10
Biological Activity Determined by the dose-dependent stimulation of thymidine uptake by BaF3 cells expressing FGF receptors. The expected ED50 is ≤ 0.5 ng/ml corresponding to a specific activity of ≥ 2 x 10^6 units/mg
Conjugate Unconjugated
Recombinant Recombinant
Sequence MLGQDMVSPE ATNSSSSSFS SPSSAGRHVR SYNHLQGDVR WRKLFSFTKY FLKIEKNGKV SGTKKENCPY SILEITSVEI GVVAVKAINS NYYLAMNKKG KLYGSKEFNN DCKLKERIEE NGYNTYASFN WQHNGRQMYV ALNGKGAPRR GQKTRRKNTS AHFLPMVVHS
Content And Storage -20°C
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 95% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.