Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Human FGF-17 Recombinant Protein, PeproTech®

Catalog No. 10027250UG
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
2 x 250 μg
25 μg
250 μg
4 x 250 μg
5 μg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
10027250UG 250 μg
10027100UG 100 μg
10-027-01M 4 x 250 μg
10-027-500 2 x 250 μg
100-27-25UG 25 μg
100-27-5UG 5 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 10027250UG Supplier Gibco™ Supplier No. 10027250UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

100-27-1MG will be provided as 4 x 250 μg (100-27-250UG). 100-27-500UG will be provided as 2 x 250 μg (100-27-250UG). Recombinant Human FGF-17 is a 22.7 kDa protein consisting of 195 amino acid residues. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

The protein encoded by this gene is a member of the fibroblast growth factor (FGF) family. FGF family members possess broad mitogenic and cell survival activities, and are involved in a variety of biological processes including embryonic development cell growth, morphogenesis, tissue repair, tumor growth and invasion. This gene was shown to be prominently expressed in the cerebellum and cortex. The mouse homolog of this gene was localized to specific sites in the midline structures of the forebrain, the midbrain-hindbrain junction, developing skeleton and developing arteries, which suggests a role in central nervous system, bone and vascular development. This gene was referred to as FGF-13 in reference 2, however, its amino acid sequence and chromosomal localization are identical to FGF17.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number O60258
For Use With (Application) ELISA, Functional Assay, Western Blot
Formulation protein with no preservative
Gene ID (Entrez) 8822
Molecular Weight (g/mol) 22.7 kDa
Name Human FGF-17
Quantity 250 μg
Source E. coli
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias FGF; FGF-13; FGF17; FGF-17; Fibroblast growth factor; Fibroblast growth factor 17; HH20; UNQ161/PRO187
Common Name FGF17
Gene Symbol FGF17
Biological Activity Determined by a cell proliferation assay using Balb/c 3T3 cells. The expected ED50 is ≤ 10 ng/ml, corresponding to a specific activity of ≥ 1 x 10^5 units/mg
Conjugate Unconjugated
Recombinant Recombinant
Sequence MTQGENHPSP NFNQYVRDQG AMTDQLSRRQ IREYQLYSRT SGKHVQVTGR RISATAEDGN KFAKLIVETD TFGSRVRIKG AESEKYICMN KRGKLIGKPS GKSKDCVFTE IVLENNYTAF QNARHEGWFM AFTRQGRPRQ ASRSRQNQRE AHFIKRLYQG QLPFPNHAEK QKQFEFVGSA PTRRTKRTRR PQPLT
Content And Storage -20°C
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 95% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.