Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Human FGF-20 Recombinant Protein, PeproTech®

Catalog No. 1004115UG
Encompass_Preferred
Change view
Click to view available options
Quantity:
15 μg
2 x 500 μg
250 μg
3 μg
500 μg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
100-41-15UG 15 μg
10-041-01M 2 x 500 μg
10041250UG 250 μg
100-41-3UG 3 μg
10041500UG 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 100-41-15UG Supplier Gibco™ Supplier No. 1004115UG
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

100-41-1MG will be provided as 2 x 500 μg (100-41-500UG). Recombinant Human FGF-20 is a 23.2 kDa protein containing 209 amino acid residues. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

FGF-20 is a secreted, heparin-binding growth factor that is a member of the FGF family. Proteins of this family play a central role during prenatal development, postnatal growth and regeneration of a variety of tissues, by promoting cellular proliferation and differentiation. FGF-20 signals through FGFR 2c and 3c, and is expressed during limb and brain development.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9NP95
For Use With (Application) Functional Assay
Formulation protein with no preservative
Gene ID (Entrez) 26281
Molecular Weight (g/mol) 23.2 kDa
Name Human FGF-20
Quantity 15 μg
Source E. coli
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias FGF; Fgf20; FGF-20; Fibroblast growth factor; Fibroblast growth factor 20; RHDA2
Common Name FGF20
Gene Symbol FGF20
Biological Activity Determined by a cell proliferation assay using Balb/c 3T3 cells. The expected ED50 is ≤ 5 ng/ml in the presence of 1 ug/ml heparin, corresponding to a specific activity of ≥ 2 x 10^5 units/mg
Conjugate Unconjugated
Recombinant Recombinant
Sequence PLAEVGGFLG GLEGLGQQVG SHFLLPPAGE RPPLLGERRS AAERSARGGP GAAQLAHLHG ILRRRQLYCR TGFHLQILPD GSVQGTRQDH SLFGILEFIS VAVGLVSIRG VDSGLYLGMN DKGELYGSEK LTSECIFREQ FEENWYNTYS SNIYKHGDTG RRYFVALNKD GTPRDGARSK RHQKFTHFLP RPVDPERVPE LYKDLLMYT
Content And Storage -20°C
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 90% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.