Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human FGF-23 ELISA Kit

Catalog No. EH189RBX10
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 x 96 Tests
5 x 96 Tests
96 Tests
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
EH189RBX10 10 x 96 Tests
EH189RBX5 5 x 96 Tests
EH189RB 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. EH189RBX10 Supplier Invitrogen™ Supplier No. EH189RBX10
Add to Cart
Add to Cart
Request a Quote

ELISA

Human FGF-23 quantitates human FGF-23 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human FGF-23.

The protein encoded by this gene is a member of the fibroblast growth factor family. FGF family members possess broad mitogenic and cell survival activities and are involved in a variety of biological processes including embryonic development, cell growth, morphogenesis, tissue repair, tumor growth and invasion. The product of this gene inhibits renal tubular phosphate transport. This gene was identified by its mutations associated with autosomal dominant hypophosphatemic rickets, an inherited phosphate wasting disorder. Abnormally high level expression of this gene was found in oncogenic hypophosphatemic osteomalacia, a phenotypically similar disease caused by abnormal phosphate metabolism. Mutations in this gene have also been shown to cause familial tumoral calcinosis with hyperphosphatemia.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number ADHR; FGF; Fgf23; FGF-23; FGFN; Fibroblast growth factor; Fibroblast growth factor 23; Fibroblast growth factor 23 C-terminal peptide; Fibroblast growth factor 23 N-terminal peptide; HPDR2; HYPF; Phosphatonin; PHPTC; tumor-derived hypophosphatemia inducing factor; Tumor-derived hypophosphatemia-inducing factor; UNQ3027/PRO9828
Assay Range 0.3 to 75 ng/mL
Assay Sensitivity 0.3 ng/mL
Conjugate HRP
Product Type ELISA
Sample Type Plasma, Serum, Supernatant
For Use With (Equipment) Colorimetric Microplate Reader
Gene ID (Entrez) 8074
Gene Symbol FGF23
Interassay CV <12%
Intraassay CV <10
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Quantity 10 x 96 Tests
Regulatory Status RUO
Sample Volume Plasma 50 μL, Serum 50 μL, Supernatant 50 μL
Storage Requirements 2°C to 8°C
Target FGF23
Test Time 1 hrs 20 mins
Total Assay Time 4 hrs 45 mins
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.