Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Human FGF-5 Recombinant Protein, PeproTech®

Catalog No. 10034500UG
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
10 μg
100 μg
50 μg
500 μg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
10034500UG 500 μg
10034100UG 100 μg
100-34-10UG 10 μg
100-34-1MG 1 mg
100-34-50UG 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 10034500UG Supplier Gibco™ Supplier No. 10034500UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Recombinant Protein

Recombinant Human FGF-5 is a 27.6 kDa protein consisting of 252 amino acid residues. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

Plays an important role in the regulation of cell proliferation and cell differentiation. Required for normal regulation of the hair growth cycle. Functions as an inhibitor of hair elongation by promoting progression from anagen, the growth phase of the hair follicle, into catagen the apoptosis-induced regression phase.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P12034
For Use With (Application) ELISA, Functional Assay, Western Blot
Formulation protein with no preservative
Gene ID (Entrez) 2250
Molecular Weight (g/mol) 27.6 kDa
Name Human FGF-5
Quantity 500 μg
Source E. coli
Regulatory Status RUO
Endotoxin Concentration <1 EU/ μg
Gene Alias angora; FGF; FGF5; FGF-5; Fibroblast growth factor; Fibroblast growth factor 5; fibroblast growth factor FGF-5; go; HBGF-5; Heparin-binding growth factor 5; Smag-82; TCMGLY
Common Name FGF5
Gene Symbol FGF5
Biological Activity Determined by dose-dependent ability to reduce tetrazolium salt, WST-8, by dehydrogenase activities of BaF3 cells expressing FGF receptors using Cell Counting Kit-8 (CCK-8). The expected ED50 is ≤ 0.5 ng/ml, corresponding to a specific activity of ≥ 2 x 10^6 units/mg
Conjugate Unconjugated
Recombinant Recombinant
Sequence MAWAHGEKRL APKGQPGPAA TDRNPIGSSS RQSSSSAMSS SSASSSPAAS LGSQGSGLEQ SSFQWSPSGR RTGSLYCRVG IGFHLQIYPD GKVNGSHEAN MLSVLEIFAV SQGIVGIRGV FSNKFLAMSK KGKLHASAKF TDDCKFRERF QENSYNTYAS AIHRTEKTGR EWYVALNKRG KAKRGCSPRV KPQHISTHFL PRFKQSEQPE LSFTVTVPEK KNPPSPIKSK IPLSAPRKNT NSVKYRLKFR FG
Content And Storage -20°C
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 95% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.