Learn More
Gibco™ Human FGF-6 Recombinant Protein, PeproTech®
Description
Recombinant Human FGF-6 is an 18.7 kDa protein consisting of 168 amino acid residues. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis
Specifications
Specifications
| Accession Number | P10767 |
| For Use With (Application) | Functional Assay |
| Formulation | protein with no preservative |
| Gene ID (Entrez) | 2251 |
| Molecular Weight (g/mol) | 18.7 kDa |
| Name | Human FGF-6 |
| Quantity | 250 μg |
| Source | E. coli |
| Regulatory Status | RUO |
| Endotoxin Concentration | <1 EU/ μg |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.