missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human FGF-7 (KGF), Animal-Free Recombinant Protein, PeproTech®

Catalog No. AF10019100
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
10 μg
100 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF10019100 100 μg
AF1001910UG 10 μg
AF100191MG 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AF10019100 Supplier Gibco™ Supplier No. AF10019100UG
Only null left
Add to Cart
Edge
Add to Cart

Recombinant Protein

Recombinant Human KGF (FGF-7) is an 18.9 kDa protein consisting of 163 amino acid residues. This product is shipped at ambient temperature. For storage, handling and reconstitution information, please see the lot-specific Certificate of Analysis

Plays an important role in the regulation of embryonic development, cell proliferation and cell differentiation. Required for normal branching morphogenesis. Growth factor active on keratinocytes. Possible major paracrine effector of normal epithelial cell proliferation.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P21781
For Use With (Application) Functional Assay
Formulation protein with no preservative
Gene ID (Entrez) 2252
Molecular Weight (g/mol) 18.9 kDa
Name Human FGF-7 (KGF), Animal-Free
Quantity 100 μg
Source E. coli
Regulatory Status RUO
Endotoxin Concentration <0.1 EU/ μg
Gene Alias Fgf7; FGF-7; Fibroblast growth factor 7; HBGF-7; Heparin-binding growth factor 7; keratinocyte growth factor; Kgf
Common Name KGF
Gene Symbol FGF7
Biological Activity Determined by a cell proliferation assay using 4 MBr-5 cells. The expected ED50 for this effect is 0.1-15 ng/ml
Conjugate Unconjugated
Recombinant Recombinant
Sequence CNDMTPEQMA TNVNCSSPER HTRSYDYMEG GDIRVRRLFC RTQWYLRIDK RGKVKGTQEM KNNYNIMEIR TVAVGIVAIK GVESEFYLAM NKEGKLYAKK ECNEDCNFKE LILENHYNTY ASAKWTHNGG EMFVALNQKG IPVRGKKTKK EQKTAHFLPM AIT
Content And Storage -20°C
Expression System E. coli
Form Lyophilized
Purity or Quality Grade ≥ 95% by SDS-PAGE gel and HPLC analyses.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.